2-Bromo-4-methoxyphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-4-methoxybenzeneboronic acid

OR72183-1G OR72183-5G
77411-090EA 619.44 USD
77411-090 77411-092
2-Bromo-4-methoxyphenylboronic acid
2-Bromo-4-methoxyphenylboronic acid
Formula: C₇H₈BBrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28142623
CAS Number: 1879166-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR