2,5-Bis(phenylthio)thiophene

Supplier: APOLLO SCIENTIFIC
Warning

OR72113-100G OR72113-50G
77411-074EA 3576.04 USD
77411-074 77411-076
2,5-Bis(phenylthio)thiophene
2,5-Bis(phenylthio)thiophene
Formula: C₁₆H₁₂S₃
MW: 300.46 g/mol
Melting Pt: 42…43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02065045
CAS Number: 2974-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR