5-Amino-2-methylquinoline hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR72088-1G OR72088-5G
77411-066EA 93.74 USD
77411-066 77411-068
5-Amino-2-methylquinoline hydrochloride
5-Amino-2-methylquinoline hydrochloride
Formula: C₁₀H₁₀N₂·ClH
CAS Number: 2828564-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR