Methyl 2-N-methylamino-6-bromobenzoate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-bromo-6-(methylamino)benzoate

OR71109-1G OR71109-250MG OR71109-5G
77411-034EA 496.16 USD
77411-034 77411-036 77411-038
Methyl 2-N-methylamino-6-bromobenzoate 95%
Methyl 2-N-methylamino-6-bromobenzoate
Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
MDL Number: MFCD34181145
CAS Number: 2654087-90-6
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR