(3-(1-Methyl-1H-pyrrol-2-yl)pyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR71106-100MG OR71106-500MG
77411-030EA 172.18 USD
77411-030 77411-032
(3-(1-Methyl-1H-pyrrol-2-yl)pyridine 95%
(3-(1-Methyl-1H-pyrrol-2-yl)pyridine
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 281 °C
Density: 1.241
MDL Number: MFCD00468104
CAS Number: 487-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR