Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate) 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Sodium phytate, Dodecasodium (2,3,4,5,6-pentaphosphonatooxycyclohexyl) phosphate

OR71104-100G OR71104-1KG OR71104-500G
77411-024EA 63.16 USD
77411-024 77411-026 77411-028
Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate) 96%
Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate)
Formula: C₆H₆Na₁₂O₂₄P₆
MW: 923.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00082310
CAS Number: 14306-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR