2-Bromo-5-cyclopropylbenzoic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR70315-1G OR70315-250MG
77410-992EA 492.08 USD
77410-992 77410-994
2-Bromo-5-cyclopropylbenzoic acid 99%
2-Bromo-5-cyclopropylbenzoic acid
Formula: C₁₀H₉BrO₂
MDL Number: MFCD29067871
CAS Number: 1692662-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR