8-Bromoquinoline-4-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 8-Bromo-4-cyanoquinoline, 8-Bromoquinoline-4-carbonitrile

OR70310-1G OR70310-250MG
77410-988EA 619.44 USD
77410-988 77410-990
8-Bromoquinoline-4-carbonitrile 95%
8-Bromoquinoline-4-carbonitrile
Formula: C₁₀H₅BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12024578
CAS Number: 1190315-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR