7-Bromo-4-cyanoquinoline 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Bromoquinoline-4-carbonitrile

OR70307-1G OR70307-250MG
77410-984EA 619.44 USD
77410-984 77410-986
7-Bromo-4-cyanoquinoline 95%
7-Bromo-4-cyanoquinoline
Formula: C₁₀H₅BrN₂
MW: 233.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32862641
CAS Number: 1537610-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR