3,5-Dimethyl-2-nitroaniline 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR62164-1G OR62164-250MG OR62164-5G
77410-804EA 496.16 USD
77410-804 77410-806 77410-808
3,5-Dimethyl-2-nitroaniline 95%
3,5-Dimethyl-2-nitroaniline
Formula: C₈H₁₀N₂O₂
MDL Number: MFCD18824861
CAS Number: 35490-74-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR