Methyl 3-(bromomethyl)-5-methylbenzoate

Supplier: APOLLO SCIENTIFIC
Danger

OR62155-1G OR62155-5G
77410-800EA 544.04 USD
77410-800 77410-802
Methyl 3-(bromomethyl)-5-methylbenzoate
Methyl 3-(bromomethyl)-5-methylbenzoate
Formula: C₁₀H₁₁BrO₂
MW: 243.1 g/mol
MDL Number: MFCD12922547
CAS Number: 120511-79-7
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR