Methyl 3,5-bis(bromomethyl)benzoate

Supplier: APOLLO SCIENTIFIC
Danger

OR62154-1G OR62154-250MG OR62154-5G
77410-794EA 263.88 USD
77410-794 77410-796 77410-798
Methyl 3,5-bis(bromomethyl)benzoate
Methyl 3,5-bis(bromomethyl)benzoate
Formula: C₁₀H₁₀Br₂O₂
MW: 321.99 g/mol
MDL Number: MFCD00182471
CAS Number: 29333-41-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR