(1S)-(+)-2-Amino-1-phenylethan-1-ol hemihydrate

Supplier: APOLLO SCIENTIFIC
Danger

OR61461-25G OR61461-5G
77410-760EA 165.04 USD
77410-760 77410-762
(1S)-(+)-2-Amino-1-phenylethan-1-ol hemihydrate
(1S)-(+)-2-Amino-1-phenylethan-1-ol hemihydrate
Formula: 2C₈H₁₁NO·H₂O
MW: 292.37 g/mol
Storage Temperature: Freezer
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR