3-Bromo-5-chloro-2-iodobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR61187-10G OR61187-1G OR61187-5G
77410-702EA 1450.8 USD
77410-702 77410-704 77410-706
3-Bromo-5-chloro-2-iodobenzonitrile 95%
3-Bromo-5-chloro-2-iodobenzonitrile
Formula: C₇H₂BrClIN
MW: 342.3600 g/mol
MDL Number: MFCD11846071
CAS Number: 1160574-37-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR