1,10-Diaminodecane

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Decane-1,10-diamine

OR51847-100G OR51847-25G OR51847-500G
77410-166EA 41.78 USD
77410-166 77410-168 77410-170
1,10-Diaminodecane
1,10-Diaminodecane
Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 140/12mm °C
Melting Pt: 59…61 °C
MDL Number: MFCD00008151
CAS Number: 646-25-3
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR