2-Bromo-5-chloroisonicotinonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR47899-1G OR47899-250MG OR47899-5G
77409-996EA 545.06 USD
77409-996 77409-998 77410-000
2-Bromo-5-chloroisonicotinonitrile 95%
2-Bromo-5-chloroisonicotinonitrile
Formula: C₆H₂BrClN₂
MW: 217.45 g/mol
MDL Number: MFCD18261449
CAS Number: 1211537-53-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR