5-Bromo-2-(cyclopropyl)thiophene-3-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR45205-1G OR45205-250MG
77409-842EA 619.44 USD
77409-842 77409-844
5-Bromo-2-(cyclopropyl)thiophene-3-carbonitrile 95%
5-Bromo-2-(cyclopropyl)thiophene-3-carbonitrile
Formula: C₈H₆BrNS
MW: 228.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32219467
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR