2-(Cyclopropyl)thiophene-3-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR45204-1G OR45204-250MG OR45204-5G
77409-836EA 495.16 USD
77409-836 77409-838 77409-840
2-(Cyclopropyl)thiophene-3-carbonitrile 95%
2-(Cyclopropyl)thiophene-3-carbonitrile
Formula: C₈H₇NS
MW: 149.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32219466
CAS Number: 2654087-86-0
UN: 1993

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR