2-Bromo-5-iodo-3-methylbenzyl alcohol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Bromo-5-iodo-3-methylphenyl)methanol

OR45195-1G OR45195-250MG OR45195-5G
77409-826EA 536.92 USD
77409-826 77409-828 77409-830
2-Bromo-5-iodo-3-methylbenzyl alcohol 95%
2-Bromo-5-iodo-3-methylbenzyl alcohol
Formula: C₈H₈BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD32868532
CAS Number: 2091676-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR