2-Bromo-4-chloro-5-iodoanisole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR400853-1G
OR400853-5G
77409-490EA
274.06
USD
77409-490
77409-492
2-Bromo-4-chloro-5-iodoanisole
2-Bromo-4-chloro-5-iodoanisole
Formula:
C₇H₅BrClIO Melting Pt: 80…83 °C |
MDL Number:
MFCD11845921 CAS Number: 1160574-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...