2-Chloro-4-methoxyphenacyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR400549-1G OR400549-250MG OR400549-5G
77409-468EA 496.16 USD
77409-468 77409-470 77409-472
2-Chloro-4-methoxyphenacyl bromide 95%
2-Chloro-4-methoxyphenacyl bromide
Formula: C₉H₈BrClO₂
MW: 263.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12139079
CAS Number: 30095-51-3
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR