3-Bromo-5-chlorobenzyl bromide 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR400325-1G OR400325-250MG OR400325-5G
77409-456EA 284.24 USD
77409-456 77409-458 77409-460
3-Bromo-5-chlorobenzyl bromide 96%
3-Bromo-5-chlorobenzyl bromide
Formula: C₇H₅Br₂Cl
MW: 284.38 g/mol
MDL Number: MFCD09261035
CAS Number: 762292-63-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR