3-Bromothiophene-4-sulphonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR315853-1G OR315853-250MG
77409-098EA 619.44 USD
77409-098 77409-100
3-Bromothiophene-4-sulphonyl chloride 95%
3-Bromothiophene-4-sulphonyl chloride
Formula: C₄H₂BrClO₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD04115406
CAS Number: 111283-90-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR