3-Chloro-4-nitroaniline 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Amino-2-chloronitrobenzene

OR2640-100G OR2640-25G OR2640-5G
77407-776EA 247.58 USD
77407-776 77407-778 77407-780
3-Chloro-4-nitroaniline 97%
3-Chloro-4-nitroaniline

Delivery of this item to mainland UK is £40, additional charge will be added at order confirmation stage by our sales team.

Formula: C6H5ClN2O2
MW: 172.57 g/mol
Melting Pt: 157…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085922
CAS Number: 825-41-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR