4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR24684-100MG OR24684-250MG
77407-530EA 21.26 USD
77407-530 77407-532
4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one 97%
4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one
Formula: C₇H₁₂N₄OS
MDL Number: MFCD00225752
CAS Number: 33509-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR