7-Methyl-4-indanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Methyl-2,3-dihydro-1H-inden-4-ol, 7-Methylindan-4-ol

OR21818-10G OR21818-1G OR21818-5G
77406-718EA 639.82 USD
77406-718 77406-720 77406-722
7-Methyl-4-indanol
7-Methyl-4-indanol
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00173675
CAS Number: 16400-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR