5-Chloro-2-nitrotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-2-methylnitrobenzene

OR17862-25G OR17862-5G
77406-128EA 28.52 USD
77406-128 77406-130
5-Chloro-2-nitrotoluene 98%
5-Chloro-2-nitrotoluene
Formula: C₇H₆ClNO₂
Melting Pt: 27…30 °C
MDL Number: MFCD00024579
CAS Number: 5367-28-2
UN: 3457
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR