2-Methoxypropene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene

OR17619-100G OR17619-25G OR17619-5G
77406-064EA 83.54 USD
77406-064 77406-066 77406-068
2-Methoxypropene
2-Methoxypropene

Delivery of this item to mainland UK is £40, additional charge will be added at order confirmation stage by our sales team.

Formula: C₄H₈O
MW: 72.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00014929
CAS Number: 116-11-0
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR