cis-2,6-Dimethylmorpholine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR16456-1G OR16456-25G OR16456-5G
77405-696EA 21.26 USD
77405-696 77405-698 77405-700
cis-2,6-Dimethylmorpholine 97%
cis-2,6-Dimethylmorpholine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 140…142/760mm °C
Melting Pt: –85 °C
Density: 0.935
Flash Pt: 41 °C
MDL Number: MFCD00078428
CAS Number: 6485-55-8
UN: 1992
ADR: 3+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR