trans,trans-2,4-Hexadien-1-ol 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
(2E,4E)-Hexa-2,4-dien-1-ol, Trans,trans-2,4-hexadien-1-ol, (2E,4E)-1-Hydroxyhexa-2,4-diene, Sorbic alcohol, trans,trans-Hexa-2,4-dien-1-ol
OR13712-10G
OR13712-5G
77404-716EA
39.74
USD
77404-716
77404-718
trans,trans-2,4-Hexadien-1-ol 95%
trans,trans-2,4-Hexadien-1-ol
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 80/12mm °C Melting Pt: 27…33 °C Density: 0.871 Flash Pt: 72 °C |
MDL Number:
MFCD00002925 CAS Number: 17102-64-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...