2-Bromo-3-thiophenecarbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromothiophene-3-carbonitrile

OR12257-1G OR12257-250MG OR12257-5G
77404-224EA 174.22 USD
77404-224 77404-226 77404-228
2-Bromo-3-thiophenecarbonitrile 95%
2-Bromo-3-thiophenecarbonitrile
Formula: C₅H₂BrNS
MW: 188.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09064981
CAS Number: 56182-43-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR