3,5-Dimethoxyphenol

Supplier: APOLLO SCIENTIFIC
Warning

OR1170-1G OR1170-5G
77404-022EA 21.26 USD
77404-022 77404-024
3,5-Dimethoxyphenol
3,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 172…175/17mm °C
Melting Pt: 40…45 °C
Flash Pt: 78 °C
MDL Number: MFCD00008388
CAS Number: 500-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR