Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR111572-1G
OR111572-250MG
77403-820EA
495.16
USD
77403-820
77403-822
Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₁H₁₁N₃O₄ MW: 249.22 g/mol |
MDL Number:
MFCD31629940 CAS Number: 1612861-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...