Methyl 3-(2,6-dichlorophenyl)acrylate

Supplier: APOLLO SCIENTIFIC
Warning

OR111147-1G OR111147-250MG
77403-668EA 467.64 USD
77403-668 77403-670
Methyl 3-(2,6-dichlorophenyl)acrylate
Methyl 3-(2,6-dichlorophenyl)acrylate
Formula: C₁₀H₈Cl₂O₂
MW: 231.08 g/mol
MDL Number: MFCD08771574
CAS Number: 46275-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR