Ethyl-5-chloro-1H-indole-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 5-chloro-1H-indole-2-carboxylate, Ethyl 5-chloro-2-indolecarboxylate

OR10934-100G OR10934-25G
77402-982EA 167.08 USD
77402-982 77402-984
Ethyl-5-chloro-1H-indole-2-carboxylate
Ethyl-5-chloro-1H-indole-2-carboxylate
Formula: C₁₁H₁₀ClNO₂
MW: 223.66 g/mol
Melting Pt: 166…168 °C
MDL Number: MFCD00005610
CAS Number: 4792-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR