3-(Boc-amino)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Aminobenzeneboronic acid, N-BOC protected, 3-(N-Boc-amino)phenylboronic acid, (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid, 3-Boronoaniline, N-BOC protected, 3-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, 3-(Boc-amino)benzeneboronic acid

OR10346-5G OR10346-1G OR10346-25G
77402-620EA 99.84 USD
77402-620 77402-616 77402-618
3-(Boc-amino)phenylboronic acid 97%
3-(Boc-amino)phenylboronic acid
Formula: C₁₁H₁₆BNO₄
MW: 237.06 g/mol
Melting Pt: 168 °C
MDL Number: MFCD03411945
CAS Number: 380430-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR