Indole-3-carbonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1H-Indole-3-carbonitrile, 3-Cyano-1H-indole, 3-Cyanoindole

OR10194-1G OR10194-5G
77402-540EA 21.26 USD
77402-540 77402-542
Indole-3-carbonitrile
Indole-3-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 179…182 °C
MDL Number: MFCD00022717
CAS Number: 5457-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR