3-Bromo-2-chloro-5-hydrazinylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (5-Bromo-6-chloropyridin-3-yl)hydrazine

OR100001-1G OR100001-250MG
77402-382EA 496.16 USD
77402-382 77402-384
3-Bromo-2-chloro-5-hydrazinylpyridine
3-Bromo-2-chloro-5-hydrazinylpyridine
Formula: C₅H₅BrClN₃
MW: 222.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33440211
CAS Number: 2357973-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR