4-(2-(Pyrrolidin-1-yl)ethyl)piperidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0806-1G OR0806-5G
77402-258EA 137.54 USD
77402-258 77402-260
4-(2-(Pyrrolidin-1-yl)ethyl)piperidine 98%
4-(2-(Pyrrolidin-1-yl)ethyl)piperidine
Formula: C₁₁H₂₂N₂
Boiling Pt: 120/4mm °C
MDL Number: MFCD03092939
CAS Number: 14759-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR