3-Amino-5-chloro-2-hydroxypyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR019982-10G OR019982-25G OR019982-5G
77401-938EA 247.58 USD
77401-938 77401-940 77401-942
3-Amino-5-chloro-2-hydroxypyridine 97%
3-Amino-5-chloro-2-hydroxypyridine
Formula: C₅H₅ClN₂O
MW: 144.55 g/mol
MDL Number: MFCD07375031
CAS Number: 98027-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR