1-Methylnaphthalene 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: α-Methylnaphthalene

OR019113-100G OR019113-25G OR019113-500G
77401-892EA 75.4 USD
77401-892 77401-894 77401-896
1-Methylnaphthalene 96%
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 240…243 °C
Melting Pt: –22 °C
Density: 1.001
Flash Pt: 82 °C
MDL Number: MFCD00004034
CAS Number: 90-12-0
UN: 3082

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR