Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: P(t-Bu)3 Pd G2

OR019112-1G OR019112-5G
77401-888EA 55.02 USD
77401-888 77401-890
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98%
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II)
Formula: C₂₄H₃₇ClNPPd
MW: 512.41 g/mol
Melting Pt: 167…170 °C
MDL Number: MFCD21608496
CAS Number: 1375325-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR