4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) (free radical) tech

Supplier: APOLLO SCIENTIFIC
Warning

OR019107-100G OR019107-25G OR019107-5G
77401-882EA 175.54 USD
77401-882 77401-884 77401-886
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) (free radical) tech
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)
Formula: C₉H₁₉N₂O
Boiling Pt: 98/3.75mm °C
Melting Pt: 33…39 °C
Density: 0.896
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR