(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol, 2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, DL-1,2-Isopropylidenglycerol, 1,2-O-Isopropylideneglycerol, Solketal, (±)-1,2-O-Isopropylidenglycerol, DL-1,2-Isopropylideneglycerol
OR019104-500G
OR019104-100G
OR019104-25G
77401-874EA
75.4
USD
77401-874
77401-870
77401-872
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 99%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
(contains isomer)
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 188…189 °C Density: 1.064 Flash Pt: 90 °C |
MDL Number:
MFCD00063238 CAS Number: 100-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...