1-(3-Bromo-4-hydroxyphenyl)ethanone 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3'-Bromo-4'-hydroxyacetophenone, 3-Bromo-4-hydroxyacetophenone

OR019102-1G OR019102-5G
77401-866EA 58.08 USD
77401-866 77401-868
1-(3-Bromo-4-hydroxyphenyl)ethanone 97%
1-(3-Bromo-4-hydroxyphenyl)ethanone
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 113 °C
MDL Number: MFCD03208974
CAS Number: 1836-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR