Sodium tetrachloropalladate(II) 98%

Supplier: APOLLO SCIENTIFIC
Warning

IN9856-1G IN9856-5G
77401-634EA 89.66 USD
77401-634 77401-636
Sodium tetrachloropalladate(II) 98%
Sodium tetrachloropalladate(II)
Formula: Cl₄Na₂Pd
MDL Number: MFCD00003487
CAS Number: 13820-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR