Palladium 5% on alumina

Supplier: APOLLO SCIENTIFIC
Warning

IN9855-25G IN9855-5G
77401-630EA 825.24 USD
77401-630 77401-632
Palladium 5% on alumina
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR