Ruthenium(III) chloride trihydrate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ruthenium trichloride trihydrate

IN9853-10G IN9853-1G IN9853-5G
77401-624EA 296.48 USD
77401-624 77401-626 77401-628
Ruthenium(III) chloride trihydrate
Ruthenium(III) chloride trihydrate
Formula: Cl₃Ru·3H₂O
MW: 261.4748 g/mol
MDL Number: MFCD00149845
CAS Number: 13815-94-6
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR