Dichloro(p-cymene)ruthenium(II) dimer 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)

IN3047-1G IN3047-5G
77401-540EA 38.72 USD
77401-540 77401-542
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Formula: C₂₀H₂₈Cl₄Ru₂
MW: 612.38 g/mol
MDL Number: MFCD00064793
CAS Number: 52462-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR