Bis(dibenzylideneacetone)palladium 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Bis(dibenzylideneacetone)palladium(0)

IN2826-1G IN2826-5G
77401-522EA 55.02 USD
77401-522 77401-524
Bis(dibenzylideneacetone)palladium 98%
Bis(dibenzylideneacetone)palladium
Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR